Assay Details
| Gene Symbol: KIAA1462 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025148358.1 | XP_025004126.1 | 3 | 3712 | 110 | |
| XM_025148359.1 | XP_025004127.1 | 3 | 1364 | 110 | ||
| XM_025148360.1 | XP_025004128.1 | 3 | 1438 | 110 | ||
| XM_418578.6 | XP_418578.5 | 3 | 3712 | 110 | ||
| GenBank mRNA | CR405765.1 | - | 1 | 50 | 110 | |
| CR407354.1 | - | 1 | 218 | 110 | ||
More Information
Set Membership: |
|

KIAA1462
Genomic Map