Assay Details
| Gene Symbol: ATAD3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015297122.2 | XP_015152608.1 | 3 - 4 | 465 | 85 | |
| GenBank mRNA | CR405946.1 | - | 3 - 4 | 199 | 85 | |
More Information
Set Membership: |
|

ATAD3B
Genomic Map