Assay Details
| Gene Symbol: ATAD3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015297122.2 | XP_015152608.1 | 5 - 6 | 592 | 79 | |
| GenBank mRNA | CR405946.1 | - | 5 - 6 | 326 | 79 | |
More Information
Set Membership: |
|

ATAD3B
Genomic Map