Assay Details
| Gene Symbol: LRRC8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642283.4 | XP_003642331.1 | 3 | 1752 | 70 | |
| XM_015279567.2 | XP_015135053.1 | 3 | 1843 | 70 | ||
| XM_015279568.2 | XP_015135054.1 | 3 | 1765 | 70 | ||
| XM_015279569.2 | XP_015135055.1 | 2 | 1699 | 70 | ||
| GenBank mRNA | CR406277.1 | - | 1 | 951 | 70 | |
More Information
Set Membership: |
|

LRRC8A
Genomic Map