Assay Details
| Gene Symbol: C1H21ORF62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CR406509.1 | - | 1 - 2 | 287 | 83 | |
| Gene Symbol: PAXBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_416707.6 | XP_416707.3 | 1 - 2 | 83 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_416707.6 transcripts.
Set Membership: |
|

C1H21ORF62
Genomic Map