Assay Details
| Gene Symbol: IQCC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282331.1 | NP_001269260.1 | 3 - 4 | 577 | 71 | |
| GenBank mRNA | BU304734.1 | - | 1 - 2 | 162 | 71 | |
| BU382037.1 | - | 3 - 4 | 605 | 71 | ||
| CR406781.1 | - | 3 - 4 | 606 | 71 | ||
More Information
Set Membership: |
|

IQCC
Genomic Map