Assay Details
| Gene Symbol: INSYN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293207.1 | NP_001280136.1 | 2 - 3 | 800 | 74 | |
| GenBank mRNA | CR407172.1 | - | 1 - 2 | 341 | 74 | |
More Information
Set Membership: |
|

INSYN1
Genomic Map