Assay Details
| Gene Symbol: TUBG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015299641.2 | XP_015155127.1 | 4 - 5 | 531 | 61 | |
| XM_025144076.1 | XP_024999844.1 | 4 - 5 | 492 | 61 | ||
| GenBank mRNA | BX929841.2 | - | 4 - 5 | 488 | 61 | |
| CR523181.1 | - | 4 - 5 | 493 | 61 | ||
More Information
Set Membership: |
|

TUBG1
Genomic Map