Assay Details
| Gene Symbol: MESDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015292034.2 | XP_015147520.1 | 1 | 1066 | 96 | |
| GenBank mRNA | CR524046.1 | - | 1 | 352 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MESDC1| Gene Symbol: MESDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015292034.2 | XP_015147520.1 | 1 | 1066 | 96 | |
| GenBank mRNA | CR524046.1 | - | 1 | 352 | 96 | |
Set Membership: |
|