Assay Details
| Gene Symbol: IRF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015299039.2 | XP_015154525.1 | 337 | 61 | ||
| XM_025143813.1 | XP_024999581.1 | 304 | 61 | |||
| XM_025143814.1 | XP_024999582.1 | 337 | 61 | |||
| XM_417990.6 | XP_417990.4 | 211 | 61 | |||
| GenBank mRNA | DQ250733.1 | - | 166 | 61 | ||
More Information
Set Membership: |
|

IRF6
Genomic Map