Assay Details
| Gene Symbol: RPL26L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277098.2 | NP_001264027.1 | 2 - 3 | 409 | 65 | |
| NM_001277099.1 | NP_001264028.1 | 2 - 3 | 211 | 65 | ||
| XM_025154820.1 | XP_025010588.1 | 2 - 3 | 295 | 65 | ||
| GenBank mRNA | BU299959.1 | - | 2 - 3 | 192 | 65 | |
| BX270382.4 | - | 1 - 2 | 169 | 65 | ||
| BX930720.1 | - | 1 - 2 | 489 | 65 | ||
| BX930842.2 | - | 2 - 3 | 408 | 65 | ||
| BX930904.2 | - | 2 - 3 | 393 | 65 | ||
| BX931915.1 | - | 2 - 3 | 409 | 65 | ||
| BX950296.2 | - | 1 - 2 | 876 | 65 | ||
| CR338863.1 | - | 2 - 3 | 197 | 65 | ||
| CR352505.1 | - | 2 - 3 | 202 | 65 | ||
| L34018.1 | - | 1 - 2 | 117 | 65 | ||
More Information
Set Membership: |
|

RPL26L1
Genomic Map