Assay Details
| Gene Symbol: RPL26L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277098.2 | NP_001264027.1 | 3 - 4 | 551 | 61 | |
| NM_001277099.1 | NP_001264028.1 | 3 - 4 | 353 | 61 | ||
| XM_025154820.1 | XP_025010588.1 | 3 - 4 | 437 | 61 | ||
| GenBank mRNA | BU299959.1 | - | 3 - 4 | 334 | 61 | |
| BX270382.4 | - | 2 - 3 | 311 | 61 | ||
| BX930720.1 | - | 2 - 3 | 631 | 61 | ||
| BX930842.2 | - | 3 - 4 | 550 | 61 | ||
| BX930904.2 | - | 3 - 4 | 535 | 61 | ||
| BX931915.1 | - | 3 - 4 | 551 | 61 | ||
| BX950296.2 | - | 2 - 3 | 1018 | 61 | ||
| CR338863.1 | - | 3 - 4 | 339 | 61 | ||
| CR352505.1 | - | 3 - 4 | 344 | 61 | ||
| L34018.1 | - | 2 - 3 | 259 | 61 | ||
More Information
Set Membership: |
|

RPL26L1
Genomic Map