Assay Details
| Gene Symbol: REL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167726.1 | NP_001161198.1 | 2 - 3 | 253 | 147 | |
| XM_015283288.2 | XP_015138774.2 | 2 - 3 | 171 | 147 | ||
| GenBank mRNA | AJ445799.1 | - | 2 - 3 | 254 | 147 | |
| BX932798.2 | - | 2 - 3 | 235 | 147 | ||
| M26381.1 | - | 1 - 2 | 132 | 147 | ||
More Information
Set Membership: |
|

REL
Genomic Map