Assay Details
| Gene Symbol: REL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167726.1 | NP_001161198.1 | 5 - 6 | 642 | 74 | |
| XM_015283288.2 | XP_015138774.2 | 5 - 6 | 560 | 74 | ||
| GenBank mRNA | AJ445799.1 | - | 5 - 6 | 643 | 74 | |
| M26381.1 | - | 4 - 5 | 521 | 74 | ||
More Information
Set Membership: |
|

REL
Genomic Map