Assay Details
| Gene Symbol: TEAD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298756.2 | XP_015154242.1 | 65 | |||
| XM_015298757.2 | XP_015154243.1 | 65 | ||||
| XM_015298758.2 | XP_015154244.1 | 65 | ||||
| XM_015298759.2 | XP_015154245.1 | 65 | ||||
| GenBank mRNA | U46127.1 | - | 1190 | 65 | ||
| U46128.2 | - | 1186 | 65 | |||
More Information
Set Membership: |
|

TEAD3
Genomic Map