Assay Details
| Gene Symbol: NDUFB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277525.1 | NP_001264454.1 | 2 - 3 | 354 | 77 | |
| GenBank mRNA | BX930426.1 | - | 2 - 3 | 354 | 77 | |
More Information
Set Membership: |
|

NDUFB6
Genomic Map