Assay Details
| Gene Symbol: GSTK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198649.1 | NP_001185578.1 | 2 - 3 | 206 | 59 | |
| XM_025149199.1 | XP_025004967.1 | 2 - 3 | 238 | 59 | ||
| GenBank mRNA | BX930797.1 | - | 2 - 3 | 170 | 59 | |
| BX931011.2 | - | 2 - 3 | 206 | 59 | ||
| BX934516.2 | - | 2 - 3 | 172 | 59 | ||
| CR387708.1 | - | 2 - 3 | 160 | 59 | ||
More Information
Set Membership: |
|

GSTK1
Genomic Map