Assay Details
| Gene Symbol: PGBD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015284385.2 | XP_015139871.1 | 3 - 4 | 2778 | 70 | |
| GenBank mRNA | BX931270.1 | - | 2 - 3 | 370 | 70 | |
More Information
Set Membership: |
|

PGBD5
Genomic Map