Assay Details
| Gene Symbol: RPRML | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025144186.1 | XP_024999954.1 | 1 | 485 | 141 | |
| GenBank mRNA | BX929633.1 | - | 1 | 436 | 141 | |
| BX931569.1 | - | 1 | 448 | 141 | ||
More Information
Set Membership: |
|

RPRML
Genomic Map