Assay Details
| Gene Symbol: FAM64A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025142121.1 | XP_024997889.1 | 3 - 4 | 684 | 79 | |
| GenBank mRNA | BX931686.1 | - | 1 - 2 | 223 | 79 | |
More Information
Set Membership: |
|

FAM64A
Genomic Map