Assay Details
| Gene Symbol: SH3BGRL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_419869.6 | XP_419869.5 | 2 - 3 | 623 | 71 | |
| GenBank mRNA | BX931923.1 | - | 1 - 2 | 234 | 71 | |
More Information
Set Membership: |
|

SH3BGRL2
Genomic Map