Assay Details
| Gene Symbol: TRAPPC3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_419775.6 | XP_419775.1 | 1 - 2 | 192 | 67 | |
| GenBank mRNA | BX932116.1 | - | 1 - 2 | 191 | 67 | |
More Information
Set Membership: |
|

TRAPPC3L
Genomic Map