Assay Details
| Gene Symbol: ORC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277719.1 | NP_001264648.1 | 3 - 4 | 393 | 65 | |
| NM_001277720.1 | NP_001264649.1 | 3 - 4 | 393 | 65 | ||
| GenBank mRNA | BU244351.1 | - | 3 - 4 | 369 | 65 | |
| BX932635.1 | - | 3 - 4 | 312 | 65 | ||
| CR385961.1 | - | 3 - 4 | 401 | 65 | ||
More Information
Set Membership: |
|

ORC6
Genomic Map