Assay Details
| Gene Symbol: GNA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415686.6 | XP_415686.1 | 1 - 2 | 342 | 64 | |
| GenBank mRNA | BX933165.1 | - | 1 - 2 | 288 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GNA13| Gene Symbol: GNA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415686.6 | XP_415686.1 | 1 - 2 | 342 | 64 | |
| GenBank mRNA | BX933165.1 | - | 1 - 2 | 288 | 64 | |
Set Membership: |
|