Assay Details
| Gene Symbol: GNA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415686.6 | XP_415686.1 | 3 - 4 | 619 | 113 | |
| GenBank mRNA | BX933165.1 | - | 3 - 4 | 565 | 113 | |
More Information
Set Membership: |
|

GNA13
Genomic Map