Assay Details
| Gene Symbol: SYBU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015283014.2 | XP_015138500.1 | 3 - 4 | 530 | 77 | |
| XM_015283015.2 | XP_015138501.1 | 3 - 4 | 533 | 77 | ||
| XM_015283016.2 | XP_015138502.1 | 3 - 4 | 523 | 77 | ||
| XM_025148137.1 | XP_025003905.1 | 3 - 4 | 545 | 77 | ||
| XM_025148138.1 | XP_025003906.1 | 2 - 3 | 444 | 77 | ||
| GenBank mRNA | BX934186.2 | - | 3 - 4 | 423 | 77 | |
| BX934240.2 | - | 3 - 4 | 423 | 77 | ||
| BX935784.1 | - | 2 - 3 | 393 | 77 | ||
More Information
Set Membership: |
|

SYBU
Genomic Map