Assay Details
| Gene Symbol: ZFAND6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277522.1 | NP_001264451.1 | 5 - 6 | 670 | 60 | |
| NM_001277526.1 | NP_001264455.1 | 5 - 6 | 611 | 60 | ||
| NM_001277528.1 | NP_001264457.1 | 4 - 5 | 462 | 60 | ||
| XM_015291998.2 | XP_015147484.1 | 5 - 6 | 580 | 60 | ||
| XM_025153839.1 | XP_025009607.1 | 5 - 6 | 497 | 60 | ||
| XM_025153840.1 | XP_025009608.1 | 4 - 5 | 405 | 60 | ||
| GenBank mRNA | BU109075.1 | - | 5 - 6 | 623 | 60 | |
| BU137192.1 | - | 4 - 5 | 412 | 60 | ||
| BX935855.1 | - | 5 - 6 | 545 | 60 | ||
| CF251301.1 | - | 4 - 5 | 464 | 60 | ||
| CN237237.1 | - | 5 - 6 | 671 | 60 | ||
| DR411810.1 | - | 5 - 6 | 609 | 60 | ||
More Information
Set Membership: |
|

ZFAND6
Genomic Map