Assay Details
| Gene Symbol: NPTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293209.1 | NP_001280138.1 | 4 - 5 | 837 | 58 | |
| XM_015278826.2 | XP_015134312.1 | 3 - 4 | 519 | 58 | ||
| XM_025153812.1 | XP_025009580.1 | 3 - 4 | 502 | 58 | ||
| GenBank mRNA | BX935877.1 | - | 2 - 3 | 122 | 58 | |
More Information
Set Membership: |
|

NPTN
Genomic Map