Assay Details
| Gene Symbol: RABL2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278112.1 | NP_001265041.1 | 6 - 7 | 548 | 96 | |
| XM_025151044.1 | XP_025006812.1 | 4 - 5 | 517 | 96 | ||
| GenBank mRNA | BX930967.1 | - | 6 - 7 | 528 | 96 | |
| BX950545.1 | - | 2 - 3 | 591 | 96 | ||
More Information
Set Membership: |
|

RABL2A
Genomic Map