Assay Details
| Gene Symbol: CDCA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276338.1 | NP_001263267.1 | 3 - 4 | 320 | 64 | |
| GenBank mRNA | BU204597.1 | - | 2 - 3 | 89 | 64 | |
| BU225566.1 | - | 3 - 4 | 320 | 64 | ||
| BU328206.1 | - | 3 - 4 | 270 | 64 | ||
| BU328800.1 | - | 3 - 4 | 269 | 64 | ||
| CR352660.1 | - | 2 - 3 | 109 | 64 | ||
| CR352941.1 | - | 3 - 4 | 269 | 64 | ||
More Information
Set Membership: |
|

CDCA9
Genomic Map