Assay Details
| Gene Symbol: CDCA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276338.1 | NP_001263267.1 | 5 - 6 | 436 | 126 | |
| GenBank mRNA | BU204597.1 | - | 4 - 5 | 205 | 126 | |
| BU225566.1 | - | 5 - 6 | 436 | 126 | ||
| BU324297.1 | - | 4 - 5 | 379 | 126 | ||
| BU328206.1 | - | 5 - 6 | 386 | 126 | ||
| BU328800.1 | - | 5 - 6 | 385 | 126 | ||
| CR352660.1 | - | 4 - 5 | 225 | 126 | ||
| CR352941.1 | - | 5 - 6 | 385 | 126 | ||
More Information
Set Membership: |
|

CDCA9
Genomic Map