Assay Details
| Gene Symbol: CDCA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276338.1 | NP_001263267.1 | 8 - 9 | 741 | 135 | |
| GenBank mRNA | BU324297.1 | - | 7 - 8 | 684 | 135 | |
| BU328206.1 | - | 8 - 9 | 695 | 139 | ||
| CR352660.1 | - | 7 - 8 | 530 | 135 | ||
| CR352941.1 | - | 8 - 9 | 690 | 135 | ||
More Information
Set Membership: |
|

CDCA9
Genomic Map