Assay Details
| Gene Symbol: CDCA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276338.1 | NP_001263267.1 | 9 - 10 | 825 | 110 | |
| GenBank mRNA | CR352660.1 | - | 8 - 9 | 614 | 110 | |
| CR352941.1 | - | 9 - 10 | 774 | 110 | ||
More Information
Set Membership: |
|

CDCA9
Genomic Map