Assay Details
| Gene Symbol: PDIK1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001232837.5 | XP_001232838.1 | 1 - 2 | 76 | 89 | |
| GenBank mRNA | CR352688.1 | - | 1 - 2 | 57 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001232837.5 transcripts.
Set Membership: |
|

PDIK1L
Genomic Map