Assay Details
| Gene Symbol: PDIK1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001232837.5 | XP_001232838.1 | 2 - 3 | 374 | 101 | |
| XM_004947732.3 | XP_004947789.1 | 1 - 2 | 525 | 101 | ||
| GenBank mRNA | CR352688.1 | - | 2 - 3 | 355 | 101 | |
More Information
Set Membership: |
|

PDIK1L
Genomic Map