Assay Details
| Gene Symbol: DSEL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004935223.3 | XP_004935280.1 | 2 | 1022 | 66 | |
| GenBank mRNA | CR352851.1 | - | 1 | 643 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DSEL| Gene Symbol: DSEL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004935223.3 | XP_004935280.1 | 2 | 1022 | 66 | |
| GenBank mRNA | CR352851.1 | - | 1 | 643 | 66 | |
Set Membership: |
|