Assay Details
| Gene Symbol: SLC33A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025153776.1 | XP_025009544.1 | 5 | 2429 | 131 | |
| XM_422831.6 | XP_422831.3 | 6 | 2642 | 131 | ||
| GenBank mRNA | CR352926.1 | - | 1 | 643 | 131 | |
More Information
Set Membership: |
|

SLC33A1
Genomic Map