Assay Details
| Gene Symbol: PTER | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004939260.3 | XP_004939317.1 | 4 - 5 | 2288 | 90 | |
| XM_015281995.2 | XP_015137481.1 | 5 - 6 | 2464 | 90 | ||
| XM_015281996.2 | XP_015137482.1 | 4 - 5 | 954 | 90 | ||
| XM_025148396.1 | XP_025004164.1 | 5 - 6 | 2502 | 90 | ||
| XM_025148397.1 | XP_025004165.1 | 4 - 5 | 2417 | 90 | ||
| XM_418628.6 | XP_418628.1 | 4 - 5 | 2379 | 90 | ||
| GenBank mRNA | CR353418.1 | - | 3 - 4 | 814 | 90 | |
More Information
Set Membership: |
|

PTER
Genomic Map