Assay Details
| Gene Symbol: LOC416257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CR353963.1 | - | 1 | 550 | 117 | |
| CR406451.1 | - | 1 | 655 | 117 | ||
| Gene Symbol: LARP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015293912.2 | XP_015149398.1 | 20 | 6406 | 117 | |
| XM_015293913.2 | XP_015149399.1 | 20 | 6404 | 117 | ||
| XM_015293914.2 | XP_015149400.1 | 20 | 6053 | 117 | ||
| XM_015293915.2 | XP_015149401.1 | 18 | 4124 | 117 | ||
More Information
Set Membership: |
|

LOC416257
Genomic Map