Assay Details
Gene Symbol: DKK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015276578.2 | XP_015132064.1 | 4 - 5 | 465 | 65 | |
XM_015276579.2 | XP_015132065.1 | 3 - 4 | 2637 | 65 | ||
XM_025150026.1 | XP_025005794.1 | 4 - 5 | 469 | 65 | ||
XM_420494.6 | XP_420494.1 | 2 - 3 | 807 | 65 | ||
GenBank mRNA | CR354152.1 | - | 2 - 3 | 793 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
