Assay Details
| Gene Symbol: MAML3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015276690.2 | XP_015132176.1 | 5 | 7118 | 74 | |
| XM_015276691.2 | XP_015132177.1 | 5 | 3062 | 74 | ||
| XM_015276692.2 | XP_015132178.1 | 5 | 7115 | 74 | ||
| GenBank mRNA | BX931050.2 | - | 1 | 548 | 74 | |
More Information
Set Membership: |
|

MAML3
Genomic Map