Assay Details
| Gene Symbol: SYNGR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252278.3 | NP_001239207.3 | 1 - 2 | 189 | 137 | |
| XM_015285214.2 | XP_015140700.2 | 1 - 2 | 212 | 137 | ||
| XM_015285219.2 | XP_015140705.1 | 1 - 2 | 212 | 137 | ||
| GenBank mRNA | BX931567.2 | - | 1 - 2 | 177 | 137 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001252278.3, XM_015285214.2, XM_015285219.2 transcripts.
Set Membership: |
|

SYNGR1
Genomic Map