Assay Details
| Gene Symbol: SFT2D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_416592.6 | XP_416592.1 | 7 - 8 | 558 | 67 | |
| GenBank mRNA | BX932470.2 | - | 7 - 8 | 479 | 67 | |
More Information
Set Membership: |
|

SFT2D2
Genomic Map