Assay Details
| Gene Symbol: IGSF21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025142730.1 | XP_024998498.1 | 4 - 5 | 598 | 66 | |
| XM_025142731.1 | XP_024998499.1 | 5 - 6 | 757 | 66 | ||
| XM_025142732.1 | XP_024998500.1 | 4 - 5 | 452 | 66 | ||
| XM_025142733.1 | XP_024998501.1 | 4 - 5 | 598 | 66 | ||
| XM_025142734.1 | XP_024998502.1 | 4 - 5 | 598 | 66 | ||
| XM_417520.6 | XP_417520.3 | 4 - 5 | 584 | 66 | ||
| GenBank mRNA | BX932742.2 | - | 4 - 5 | 573 | 66 | |
More Information
Set Membership: |
|

IGSF21
Genomic Map