Assay Details
| Gene Symbol: DTX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015296006.2 | XP_015151492.2 | 3 - 4 | 1133 | 66 | |
| XM_015296007.2 | XP_015151493.2 | 3 - 4 | 1133 | 66 | ||
| XM_015296008.2 | XP_015151494.2 | 3 - 4 | 1133 | 66 | ||
| XM_015296009.2 | XP_015151495.2 | 3 - 4 | 1133 | 66 | ||
| GenBank mRNA | BX933137.2 | - | 2 - 3 | 609 | 66 | |
| CR387657.1 | - | 1 - 2 | 123 | 66 | ||
More Information
Set Membership: |
|

DTX2
Genomic Map