Assay Details
| Gene Symbol: AMPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199588.1 | NP_001186517.1 | 2 - 3 | 415 | 116 | |
| XM_015286376.2 | XP_015141862.1 | 2 - 3 | 628 | 116 | ||
| XM_015286378.2 | XP_015141864.1 | 2 - 3 | 629 | 116 | ||
| GenBank mRNA | BX929327.2 | - | 2 - 3 | 416 | 116 | |
| BX933266.2 | - | 2 - 3 | 330 | 116 | ||
More Information
Set Membership: |
|

AMPD3
Genomic Map