Assay Details
| Gene Symbol: LOC421982 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015285030.2 | XP_015140516.1 | 3 - 4 | 395 | 88 | |
| XM_015285031.2 | XP_015140517.1 | 3 - 4 | 395 | 88 | ||
| XM_419992.6 | XP_419992.1 | 3 - 4 | 395 | 88 | ||
| GenBank mRNA | BX930463.2 | - | 3 - 4 | 349 | 88 | |
| BX933584.2 | - | 3 - 4 | 372 | 88 | ||
More Information
Set Membership: |
|

LOC421982
Genomic Map