Assay Details
| Gene Symbol: C1QA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_417654.6 | XP_417654.2 | 2 - 3 | 256 | 57 | |
| GenBank mRNA | BX934534.2 | - | 2 - 3 | 222 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1QA| Gene Symbol: C1QA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_417654.6 | XP_417654.2 | 2 - 3 | 256 | 57 | |
| GenBank mRNA | BX934534.2 | - | 2 - 3 | 222 | 57 | |
Set Membership: |
|