Assay Details
| Gene Symbol: DTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199402.1 | NP_001186331.1 | 5 - 6 | 679 | 71 | |
| GenBank mRNA | BX929594.1 | - | 5 - 6 | 670 | 71 | |
| BX935203.2 | - | 4 - 5 | 432 | 71 | ||
More Information
Set Membership: |
|

DTD1
Genomic Map