Assay Details
| Gene Symbol: PDGFRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001231867.5 | XP_001231868.2 | 1 - 2 | 89 | 68 | |
| XM_004935998.3 | XP_004936055.1 | 1 - 2 | 89 | 68 | ||
| GenBank mRNA | BX935596.2 | - | 1 - 2 | 50 | 68 | |
| HG983522.1 | - | 1 - 2 | 68 | 68 | ||
More Information
Set Membership: |
|

PDGFRL
Genomic Map